Order Entry
United States
ContactUsLinkComponent
Yttrium Standard: Y @ 10 µg/mL in 2% HNO3, VHG, LGC Standards
Yttrium Standard: Y @ 10 µg/mL in 2% HNO3, VHG, LGC Standards
  77695-351
New Product
 :  
undefined
Yttrium Standard: Y @ 10 µg/mL in 2% HNO3, VHG, LGC Standards
  77695-351
 :  VHG-LYN-100
New Product
 :  

 

  • Concentration:
    10 µg/mL
  • Matrix:
    2% HNO₃
  • Description:
    Yttrium Standard: Y @ 10 µg/mL in 2% HNO3
  • Size:
    100 ml
  • Cat. No.:
    77695-351