Sulfur Standard for ASTM D5623: Multi-component standard containing 22 sulfur species @ 50 µg/g (as Component) in Base Fuel, VHG, LGC Standards
77695-353
New Product
:
- Concentration:50 µg/g
- Matrix:Base Oil 20
- Description:Sulfur Standard for ASTM D5623: Multi-component standard containing 22 sulfur species @ 50 µg/g (as Component) in Base Fuel
- Size:2 ml
- Cat. No.:77695-353