Cadmium Standard: Cd @ 1000 µg/g in Hydrocarbon Oil, VHG, LGC Standards
Catalog # 77693-183
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Concentration:1000 µg/g
- Matrix:oil
- Description:Cadmium Standard: Cd @ 1000 µg/g in Hydrocarbon Oil
- Size:50 g
- Cat. No.:77693-183