Copper Standard: Cu @ 5000 µg/g in Hydrocarbon Oil, VHG, LGC Standards
Catalog # 77693-196
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Concentration:5000 µg/g
- Matrix:oil
- Description:Copper Standard: Cu @ 5000 µg/g in Hydrocarbon Oil
- Size:50 g
- Cat. No.:77693-196