Order Entry
United States
ContactUsLinkComponent
 
Titanium Standard: Ti @ 5000 µg/g in Hydrocarbon Oil, VHG, LGC Standards
Catalog # 77695-373
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Titanium Standard: Ti @ 5000 µg/g in Hydrocarbon Oil, VHG, LGC Standards
Catalog # 77695-373
Supplier:  LGC Standards
Supplier Number:  VHG-OTI-5000-50G
New Product
CAS Number:  

Specifications

  • Concentration:
    5000 µg/g
  • Matrix:
    oil
  • Description:
    Titanium Standard: Ti @ 5000 µg/g in Hydrocarbon Oil
  • Size:
    50 g
  • Cat. No.:
    77695-373

Specifications

About this item