Order Entry
United States
ContactUsLinkComponent
Yttrium Standard: Y @ 2 wt% in Hydrocarbon Oil, VHG, LGC Standards
Yttrium Standard: Y @ 2 wt% in Hydrocarbon Oil, VHG, LGC Standards
Catalog # 77695-382
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Yttrium Standard: Y @ 2 wt% in Hydrocarbon Oil, VHG, LGC Standards
Catalog # 77695-382
Supplier:  LGC Standards
Supplier Number:  VHG-OYDN-2P-100G
New Product
CAS Number:  

Specifications

  • Concentration:
    2 wt%
  • Matrix:
    oil
  • Description:
    Yttrium Standard: Y @ 2 wt% in Hydrocarbon Oil
  • Size:
    100 g
  • Cat. No.:
    77695-382

Specifications

About this item