Zirconium Standard: Zr @ 5000 µg/g in Hydrocarbon Oil, VHG, LGC Standards
Catalog # 77695-386
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Concentration:5000 µg/g
- Matrix:oil
- Description:Zirconium Standard: Zr @ 5000 µg/g in Hydrocarbon Oil
- Size:50 g
- Cat. No.:77695-386