Sulfur Standard: S @ 2500 µg/g (0.2500 wt%) in 20 cSt Mineral Oil, VHG, LGC Standards
77695-445
New Product
:
- Concentration:2500 µg/g
- Matrix:Mineral oil
- Description:Sulfur Standard: S @ 2500 µg/g (0.2500 wt%) in 20 cSt Mineral Oil
- Size:100 ml
- Cat. No.:77695-445