Order Entry
United States
ContactUsLinkComponent
Sulfur Standard: S @ 15 ug/g in 70% Isooctane/30% Toluene, VHG, LGC Standards
Sulfur Standard: S @ 15 ug/g in 70% Isooctane/30% Toluene, VHG, LGC Standards
Catalog # 77695-524
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Sulfur Standard: S @ 15 ug/g in 70% Isooctane/30% Toluene, VHG, LGC Standards
Catalog # 77695-524
Supplier:  LGC Standards
Supplier Number:  VHG-SISO70TOL30-15-100
New Product
CAS Number:  

Specifications

  • Concentration:
    15 ug
  • Description:
    Sulfur Standard: S @ 15 ug/g in 70% Isooctane/30% Toluene
  • Size:
    100 ml
  • Cat. No.:
    77695-524

Specifications

About this item