Order Entry
United States
ContactUsLinkComponent
Multi-Elements Standard 8 Components US EPA RCRA: 100 µg/mL each of Ag, As, Ba, Cd, Cr, Hg, Pb, Se in 5% HNO3, VHG, LGC Standards
Multi-Elements Standard 8 Components US EPA RCRA: 100 µg/mL each of Ag, As, Ba, Cd, Cr, Hg, Pb, Se in 5% HNO3, VHG, LGC Standards
Catalog # 77695-558
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Multi-Elements Standard 8 Components US EPA RCRA: 100 µg/mL each of Ag, As, Ba, Cd, Cr, Hg, Pb, Se in 5% HNO3, VHG, LGC Standards
Catalog # 77695-558
Supplier:  LGC Standards
Supplier Number:  VHG-SM45-100
New Product
CAS Number:  

Specifications

  • Concentration:
    100 µg/mL
  • Matrix:
    5% HNO₃
  • Description:
    US EPA RCRA Elements Standard: Ag, As, Ba, Cd, Cr, Hg, Pb, Se @ 100 µg/mL in 5% HNO3
  • Size:
    100 ml
  • Cat. No.:
    77695-558

Specifications

About this item