Sulfur Standard: S @ 75 µg/g (0.0075 wt%) in 75 cSt Mineral Oil, VHG, LGC Standards
Catalog # 77695-585
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Concentration:75 µg/g
- Matrix:Mineral oil
- Description:Sulfur Standard: S @ 75 µg/g (0.0075 wt%) in 75 cSt Mineral Oil
- Size:100 ml
- Cat. No.:77695-585