Order Entry
United States
ContactUsLinkComponent
Beryllium Standard: Be @ 10000 µg/mL in 5% HNO3, VHG, LGC Standards
Beryllium Standard: Be @ 10000 µg/mL in 5% HNO3, VHG, LGC Standards
Catalog # 77693-288
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Beryllium Standard: Be @ 10000 µg/mL in 5% HNO3, VHG, LGC Standards
Catalog # 77693-288
Supplier:  LGC Standards
Supplier Number:  VHG-TBEN-100
New Product
CAS Number:  

Specifications

  • Concentration:
    10000 µg/mL
  • Matrix:
    5% HNO₃
  • Description:
    Beryllium Standard: Be @ 10000 µg/mL in 5% HNO3
  • Size:
    100 ml
  • Cat. No.:
    77693-288

Specifications

About this item