Order Entry
United States
ContactUsLinkComponent
 
Cerium Standard: Ce @ 10000 µg/mL in 5% HNO3, VHG, LGC Standards
  77693-299
New Product
 :  
undefined
Cerium Standard: Ce @ 10000 µg/mL in 5% HNO3, VHG, LGC Standards
  77693-299
 :  VHG-TCEN-100
New Product
 :  

 

  • Concentration:
    10000 µg/mL
  • Matrix:
    5% HNO₃
  • Description:
    Cerium Standard: Ce @ 10000 µg/mL in 5% HNO3
  • Size:
    100 ml
  • Cat. No.:
    77693-299