Order Entry
United States
ContactUsLinkComponent
Chromium Standard: Cr @ 10000 µg/mL in 5% HCl, VHG, LGC Standards
Chromium Standard: Cr @ 10000 µg/mL in 5% HCl, VHG, LGC Standards
Catalog # 77693-302
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Chromium Standard: Cr @ 10000 µg/mL in 5% HCl, VHG, LGC Standards
Catalog # 77693-302
Supplier:  LGC Standards
Supplier Number:  VHG-TCRH-100
New Product
CAS Number:  

Specifications

  • Concentration:
    10000 µg/mL
  • Matrix:
    5% HCl
  • Description:
    Chromium Standard: Cr @ 10000 µg/mL in 5% HCl
  • Size:
    100 ml
  • Cat. No.:
    77693-302

Specifications

About this item