Order Entry
United States
ContactUsLinkComponent
Zirconium Standard: Zr @ 10000 µg/mL in 5% HCl, VHG, LGC Standards
Zirconium Standard: Zr @ 10000 µg/mL in 5% HCl, VHG, LGC Standards
Catalog # 77695-644
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Zirconium Standard: Zr @ 10000 µg/mL in 5% HCl, VHG, LGC Standards
Catalog # 77695-644
Supplier:  LGC Standards
Supplier Number:  VHG-TZRH-100
New Product
CAS Number:  

Specifications

  • Concentration:
    10000 µg/mL
  • Matrix:
    5% HCl
  • Description:
    Zirconium Standard: Zr @ 10000 µg/mL in 5% HCl
  • Size:
    100 ml
  • Cat. No.:
    77695-644

Specifications

About this item