Order Entry
United States
ContactUsLinkComponent
Phosphate Buffered Saline (PBS) with 0.02% Tween® 80
Phosphate Buffered Saline (PBS) with 0.02% Tween® 80
Catalog # 77623-682
Supplier:  Hardy Diagnostics
New Product
CAS Number:  
Phosphate Buffered Saline (PBS) with 0.02% Tween® 80
Catalog # 77623-682
Supplier:  Hardy Diagnostics
Supplier Number:  U334
New Product
CAS Number:  

Specifications

  • Pack type:
    Polycarbonate container
  • Cat. No.:
    77623-682
  • pH:
    7.5 ±0.1 at 25 °C
  • Fill volume:
    50 ml

Specifications

About this item

0.01 M Phosphate Buffered Saline with 0.02% Tween® 80 (PBST) is recommended for use in performing microbial sampling, culture, and surveillance procedures for reprocessed endoscopes.