ContactUsLinkComponent
Start-up Kit, ISO-COP-2
Start-up Kit, ISO-COP-2
Catalog # 77663-984
New Product
Start-up Kit, ISO-COP-2
Catalog # 77663-984
Supplier Number:  95699
New Product
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Startup kit, 4 sleeves
  • Cat. No.:
    77663-984
Specifications
About this item
The ISO-COP-2 start-up kit contains everything needed to begin working with this sensor. This kit is highly recommended for first-time ISO-COP-2 users.
  • 10 ml electrolyte filling solution for CO sensor
Delivery: Kit includes five additional membrane sleeves (Sold individually in packages of four sleeves.), electrolyte filling solution, MicroFil™ electrolyte filling needle, 1 ml syringe, two sample vials with lids and stoppers, potentiometer adjustment tool (Tweaker).