ContactUsLinkComponent
USB Thumb Drive
USB Thumb Drive
Catalog # 77662-074
New Product
USB Thumb Drive
Catalog # 77662-074
Supplier Number:  300749
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    EVOM3 USB drive 32 GB, programmed with Python digital output package
  • Cat. No.:
    77662-074
Specifications
About this item
This USB thumb drive is loaded with the Python digital output package for use with the popular EVOM™ Manual TEER (transepithelial electrical resistance) meter.
A blank replacement thumb drive is also available. This thumb drive hold 32 Gb of data and is suitable for use with the EVOM™ Manual for auto data logging of all your measurements. The retractable design of this SanDisk thumb drive protects the USB connector when the unit is not in use.