ContactUsLinkComponent
Mobile Computer Workstation with Adjustable Shelves, Bandy
Mobile Computer Workstation with Adjustable Shelves, Bandy
Catalog # 98000-580
Supplier:  Bandy
Product available on GSA Advantage®
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
undefined
Mobile Computer Workstation with Adjustable Shelves, Bandy
Catalog # 98000-580
Supplier:  Bandy
Supplier Number:  CRT60002
Product available on GSA Advantage®
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Specifications
  • Cat. No.:
    98000-580
  • Shipping weight:
    80 lbs.
  • Supplier no.:
    CRT60002
Specifications
About this item
Workstation is made from 16-gauge, Type 304 stainless steel.
  • Completely electropolished
  • Includes four caster wheels, with brakes on two
Desktop and two adjustable shelves are perforated.
Ordering information: For optional upright CPU stand (shown), contact your Avantor sales representative.