ContactUsLinkComponent
Vacuum Gauge, Dial, Series CG16K, Edwards
Vacuum Gauge, Dial, Series CG16K, Edwards
  EVD356-10-000
 :  Edwards
undefined
Vacuum Gauge, Dial, Series CG16K, Edwards
  EVD356-10-000
 :  Edwards
 :  D35610000
  • Range:
    0–1040 mbar
  • Description:
    Vacuum Gauge
  • Cat. No.:
    EVD356-10-000
  • Supplier no.:
    D356-10-000
  • Int. Volume:
    160 mL
  • Max. Over-Pressure:
    2 atm absolute, 1 atm gauge
This gauge is barometrically independent and ruggedly constructed
Easy-to-read linear scale. Suitable for panel mounting or direct pipeline mounting via NW16 flange. Accuracy is ±2% of full scale deflection