About this item
Thixotropic, synthetic acrylic latex masking compounds to temporarily mask areas of printed circuit boards
- 100% water-soluble masks
- Excellent for protecting plated thru-holes and contact surfaces
- Masks are safe on precious metals and sensitive contacts
The peelable, water-soluble masks are designed to withstand fluxing, wave soldering, and cleaning operations. These masks are antistatic, offer excellent heat stability, and have no offensive odor.
Designed for use in closed-loop/in-line aqueous systems.
The Hi-Temp masks are formulated with noncontaminating, noncorrosive materials to withstand high temperatures. Masks exhibit very low ionic residues and are compatible with all organic and synthetic fluxes. These nonflammable, heat-stable masks are quickly removed in batch cleaning or in-line water systems. Will not harm metals.
Choose from peelable and water soluble formulations. Masks conform to MIL-STD-2000 paragraph 5.3.18, parts a, b, & d.