ContactUsLinkComponent
8577 P95 Particulate Respirator with Nuisance Level Organic Vapor Relief, 3M™
8577 P95 Particulate Respirator with Nuisance Level Organic Vapor Relief, 3M™
Catalog # 56222-366
Supplier:  3M
undefined
8577 P95 Particulate Respirator with Nuisance Level Organic Vapor Relief, 3M™
Catalog # 56222-366
Supplier:  3M
Supplier Number:  7000002062
Specifications
  • Description:
    8577 P95 Particulate respirator with nuisance level organic vapor relief
  • Cat. No.:
    56222-366
  • Supplier no.:
    8577
Specifications
About this item
Excellent choice for work settings that involve heat, humidity, and long wear periods.
  • Designed to provide comfortable, reliable worker protection against certain oil and non-oil based aerosol particles
  • Features 3M Cool Flow® Exhalation valve for cool, dry comfort
  • Adjustable "M" nose clip provides custom fit and secure seal
Designed to provide comfortable, reliable worker protection against certain oil and non-oil-based aerosol particles including those present with nuisance levels of organic vapors such as solvents, degreasers, and resins. (Nuisance level refers to concentrations not exceeding the OSHA PEL or applicable government occupational exposure limits, whichever is lower.) Features 3M Cool Flow® Exhalation valve for cool, dry comfort and “M” nose clip to help provide a custom fit and secure seal.
Suggested uses include foundry operations, lab settings, agriculture, petrochemical manufacturing, and undercoating.
Caution: All 3M™ products must be used in accordance with OSHA regulations and the user instructions, warnings, and limitations accompanying each product.