2315 Disposable N99 Particulate Respirator with Adjustable Straps, Moldex®
56219-016
Small Business Enterprise
- Description:2315 N99 Particulate Respirator with Adjustable Straps
- Environmentally Preferable:Safer Human and Environmental Health
- Cat. No.:56219-016
- Supplier no.:2315N99
Disposable respirator provides a filter efficiency level of 99% or greater against particulate aerosols free of oil up to 10 times OSHA Permissible Exposure Limit (PEL).
- Exhale valve lets worker breathe out easily, stay cool longer
- Durable mask with crush-resistant Dura-Mesh® outer shell will not absorb moisture or collapse
- Contoured facepiece features a soft foam nose seal for a comfortable fit
- Softspun® lining keeps workers comfortable and stays clean-looking longer
- Adjustable straps assure a snug, comfortable fit
Flame retardants help decrease flammability of the shell. Fits easily under welding hoods.