Top Gun™ Ionizing Air Gun, Simco Ion
Supplier: Simco
Certificates
About this item
Highly reliable ionizing air gun for electronics manufacturing applications
The 120V models are UL and C-UL listed. The 230V models are CE marked.
Balanced to ±15V, the air gun features high blow-off force to provide efficient cleaning and rapid static discharge. Uses compressed air or nitrogen. The gun body is lightweight, durable, and features a light-touch trigger. Both the gun body and the cable/hose assembly are static dissipative. Airflow control and ion balance control functions are built into the handle for easy operator use. A two-level LED indicates both power on and active ionization.
Air input: 5 - 100 psi
Discharge time: 1000 - 100 V 1.3 s at 15.2 cm (6") and 30 psi
Frequently Bought Together