Order Entry
United States
ContactUsLinkComponent
 
Deuterium Halogen Light Source With Integrated TTL Shutter, D4H
  77664-188
New Product
Deuterium Halogen Light Source With Integrated TTL Shutter, D4H
  77664-188
 :  D4H
New Product

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Description:
    D4H Deuterium halogen light source, 17.8×15.7×25 cm, 6 kg
  • Cat. No.:
    77664-188
  • Electrical:
    110/240 V, 50 - 60 Hz, 1 A

 

 

The D4H is a combined deuterium and halogen light source for UV/VIS and NIR applications. This light source is ideally suited to work with WPI’s spectrometer modules and sample cells. It supplies a continuous spectrum in the UV, VIS and NIR range from 200 to 1100 nm. The D4H is equipped with an integrated electrical shutter, which can be controlled by a switch or a TTL signal.

  • SMA fiber optic connection
  • Separate UV and VIS bulb control
  • High output: twice the intensity (compared to conventional models)
  • High stability
  • Long Life Lamp: 2000 hours
  • External control
  • Shutter function

Benefits:
Matched deuterium and halogen bulbs
Matched optical output for WPI's flow cells and fiber optic probes
Low drift <1 mAU/h @ 254 nm
Simple exchange of light bulbs

Applications:
UV/VIS/NIR absorbance spectroscopy
Excitation light source for fluorescence applications
Colored Dissolved Organic Matter (CDOM) detection in seawater
Nutrient Analysis in fresh water, drinking water and seawater using WPI's LWCCs
Protein detection using WPI's V-Vette or DIP UV MINI fiber optic probes