ContactUsLinkComponent
Universal Dipping Probe Stand
Catalog # 77662-096
New Product
Universal Dipping Probe Stand
Catalog # 77662-096
Supplier Number:  42200
New Product
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Universal dipping probe stand
  • Cat. No.:
    77662-096
Specifications
About this item
The universal dipping probe stand is designed to holds dip tips, diptip minis, spectropipetters and biosensors (macro and micro).
Delivery: Kit includes: Small V-base, 25 cm stainless steel post and holder bracket, delrin with thumbscrew