Mouse Recombinant Mouse Glioma CultuPB (from E. coli)
Catalog # 77661-236
Supplier: Fujifilm Irvine Scientific
New Product
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Mouse
- Size:500 µg
- Storage Conditions:At or below –20 °C
- Endotoxin-free:N
- Carrier-Free:Y
- Protease-free:N
- Animal-Free:N
- Protein/Peptide Name:Mouse Glioma CultuPB
- Sequence:NA
- Endotoxin Level:Low
- Nuclease-free:N
- Grade:RUO
- Shipping Temperature:RT
- Cat. No.:77661-236
Specifications
About this item
Cytokines for cell culture of mouse glioma cells. A glioma is a generic term used to describe tumors that arise from the glia tissue of the brain that supports and surrounds neurons. Glioma tumor cells can be successfully cultured ex vivo with EGF, FGF-basic (or FGF-2), and PDGFs (AA and BB).
- High quality
- Low endotoxin
- Affordable
Delivery information: 2×100 μg of Recombinant mouse EGF, 200 μg of Recombinant mouse FGF-basic/FGF-2, 100 μg of Recombinant mouse PDGF-AA and 100 μg of Recombinant mouse PDGF-BB.
Caution: For research use only.