Catherization Cannulation Kit
Product Details & Documents
Nine piece surgical instrument kit ideal for catherization and cannulation.
Delivery information: Kit includes: Disposable scalpel, #15, sterile, 10/box, lever action skin stapler 35S, sterile, Metzenbaum scissors, 11.5 cm, SuperCut, straight, castroviejo needle holder with lock, 14 cm, straight, serrated, Halsted mosquito hemostatic forceps, 12.5 cm, straight, Spence stitch ligature scissors, 9 cm, Adson-Brown forceps, 12 cm, straight, TC jaws, 7×7 teeth, WPI Swiss tweezers #5, 11 cm, 0.1×0.06 mm tips, Nugent utility forceps, 10 mm, angled tip.
Specifications
- Description:Catherization and cannulation kit
- Material:Stainless steel