Phenol-Mix 1 50 µg/mL in Methanol, Dr. Ehrenstorfer, LGC Standards
Catalog # 77679-147
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Matrix:Methanol
- Pack type:Glass bottle
- Description:Phenol-Mi x 1 50 µg/mL in Methanol
- Size:10 ml
- Cat. No.:77679-147