Order Entry
United States
ContactUsLinkComponent
Tamoxifen, TRC, LGC Standards
Tamoxifen, TRC, LGC Standards
Supplier:  LGC Standards
New Product
Certificates
undefined
Tamoxifen, TRC, LGC Standards
Supplier:  LGC Standards
New Product

Specifications

  • CAS No.:
    10540-29-1
  • Formula:
    C₂₆H₂₉NO
  • Molecular Weight:
    371.524 g/mol

Specifications

Frequently Bought Together