(S)-Mephenytoin, TRC, LGC Standards
Catalog # 77694-705
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:70989-04-7
- Description:(S)-Mephenytoin
- Size:100 mg
- Formula:C₁₂H₁₄N₂O₂
- Molecular Weight:218.3 g/mol
- Cat. No.:77694-705