Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United States
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
25-Hydroxyvitamin D3 3-Hemisuccinate, TRC, LGC Standards
25-Hydroxyvitamin D3 3-Hemisuccinate, TRC, LGC Standards
25-Hydroxyvitamin D3 3-Hemisuccinate, TRC, LGC Standards
77694-563
:
LGC Standards
New Product
:
25-Hydroxyvitamin D3 3-Hemisuccinate, TRC, LGC Standards
77694-563
:
LGC Standards
:
TRC-H995855-1MG
New Product
:
CAS No.:
69511-19-9
Description:
25-Hydroxyvitamin D3 3-Hemisuccinate
Size:
1 mg
Formula:
C₃₁H₄₈O₅
Molecular Weight:
500.7098 g/mol
Cat. No.:
77694-563