Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United States
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
1-Hydroxypyrene, TRC, LGC Standards
1-Hydroxypyrene, TRC, LGC Standards
1-Hydroxypyrene, TRC, LGC Standards
77694-551
:
LGC Standards
New Product
:
1-Hydroxypyrene, TRC, LGC Standards
77694-551
:
LGC Standards
:
TRC-H952700-100MG
New Product
:
CAS No.:
5315-79-7
Description:
1-Hydroxypyrene
Size:
100 mg
Formula:
C₁₆H₁₀O
Molecular Weight:
218.255 g/mol
Cat. No.:
77694-551