Gleevec (Imatinib Mesylate), TRC, LGC Standards
Catalog # 77694-434
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:220127-57-1
- Description:Gleevec (Imatinib Mesylate)
- Size:100 mg
- Formula:C₂₉H₃₁N₇O·CH₄SO₃
- Molecular Weight:589.72 g/mol
- Cat. No.:77694-434