Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United States
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
17beta-Estradiol-d4, TRC, LGC Standards
17beta-Estradiol-d4, TRC, LGC Standards
17beta-Estradiol-d4, TRC, LGC Standards
Catalog # 77694-343
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
17beta-Estradiol-d4, TRC, LGC Standards
Catalog # 77694-343
Supplier:
LGC Standards
Supplier Number:
TRC-E888004-1MG
New Product
CAS Number:
Specifications
CAS No.:
66789-03-5
Description:
17beta-Estradiol-d4
Size:
1 mg
Cat. No.:
77694-343
Specifications
About this item
Product Details
Product Details