7,9-Di-tert-butyl-1-oxaspiro[4.5]deca-6,9-diene-2,8-dione, TRC, LGC Standards
77694-281
New Product
:
- CAS No.:82304-66-3
- Description:7,9-Di-tert-butyl-1-oxaspiro[4.5]deca-6,9-diene-2,8-dione
- Size:100 mg
- Formula:C₁₇H₂₄O₃
- Molecular Weight:276.37 g/mol
- Cat. No.:77694-281