5-Aminoimidazole-4-carboxamide-1-Beta-D-ribofuranoside, TRC, LGC Standards
Catalog # 77693-823
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:2627-69-2
- Description:5-Aminoimidazole-4-carboxamide-1-Beta-D-ribofuranoside
- Size:50 mg
- Formula:C₉H₁₄N₄O₅
- Molecular Weight:258.23 g/mol
- Cat. No.:77693-823