N-Acetyllactosamine, TRC, LGC Standards
Catalog # 77693-784
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:32181-59-2
- Description:N-Acetyllactosamine
- Size:100 mg
- Formula:C₁₄H₂₅NO₁₁
- Molecular Weight:383.35 g/mol
- Cat. No.:77693-784