6-Demethylazithromycin (Azaerythromycin A), Mikromol, LGC Standards
Catalog # 77692-939
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:76801-85-9
- Description:6-Demethylazithromycin (Azaerythromycin A)
- Size:100 mg
- Formula:C₃₇H₇₀N₂O₁₂
- Molecular Weight:734.96 g/mol
- Cat. No.:77692-939