Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United States
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
4-Nitrosohydrochlorothiazide, Mikromol, LGC Standards
4-Nitrosohydrochlorothiazide, Mikromol, LGC Standards
4-Nitrosohydrochlorothiazide, Mikromol, LGC Standards
Catalog # 77692-479
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
4-Nitrosohydrochlorothiazide, Mikromol, LGC Standards
Catalog # 77692-479
Supplier:
LGC Standards
Supplier Number:
MM0011.12-0025
New Product
CAS Number:
Specifications
CAS No.:
63779-86-2
Description:
4-Nitrosohydrochlorothiazide
Size:
25 mg
Cat. No.:
77692-479
Specifications
About this item
Product Details
Product Details