Chlorzoxazone, LoGiCal, LGC Standards
Catalog # 77692-410
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:95-25-0
- Description:Chlorzoxazone
- Size:10 mg
- Formula:C₇H₄ClNO₂
- Molecular Weight:169.57 g/mol
- Cat. No.:77692-410