Order Entry
United States
ContactUsLinkComponent
Dexamethasone, LoGiCal, LGC Standards
Dexamethasone, LoGiCal, LGC Standards
Supplier:  LGC Standards
New Product
Certificates
undefined
Dexamethasone, LoGiCal, LGC Standards
Supplier:  LGC Standards
New Product

Specifications

  • CAS No.:
    50-02-2
  • Formula:
    C₂₂H₂₉FO₅
  • Molecular Weight:
    392.47 g/mol

Specifications

Frequently Bought Together