Chlorzoxazone 1.0 mg/ml in Acetonitrile, LoGiCal, LGC Standards
Catalog # 77692-304
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:95-25-0
- Description:Chlorzoxazone 1.0 mg/ml in Acetonitrile
- Size:1 ml
- Formula:C₇H₄ClNO₂
- Molecular Weight:169.57 g/mol
- Cat. No.:77692-304