Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United States
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
Chlorpromazine-D3 0.1 mg/ml in Methanol, LoGiCal, LGC Standards
Chlorpromazine-D3 0.1 mg/ml in Methanol, LoGiCal, LGC Standards
77692-283
:
LGC Standards
New Product
:
Chlorpromazine-D3 0.1 mg/ml in Methanol, LoGiCal, LGC Standards
77692-283
:
LGC Standards
:
LGCAMP0420.80-02
New Product
:
CAS No.:
136765-28-1
Description:
Chlorpromazine-D3 0.1 mg/ml in Methanol
Size:
1 ml
Cat. No.:
77692-283