Acetylsalicylic Acid 1.0 mg/ml in Acetonitrile, LoGiCal, LGC Standards
Catalog # 77692-264
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:50-78-2
- Description:Acetylsalicylic Acid 1.0 mg/ml in Acetonitrile
- Size:1 ml
- Formula:2-(CH₃CO₂)C₆H₄CO₂H
- Molecular Weight:180.16 g/mol
- Cat. No.:77692-264