Demoxepam 1.0 mg/ml in Methanol, LoGiCal, LGC Standards
Catalog # 77692-239
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:963-39-3
- Description:Demoxepam 1.0 mg/ml in Methanol
- Size:1 ml
- Formula:C₁₅H₁₁ClN₂O₂
- Molecular Weight:286.72 g/mol
- Cat. No.:77692-239