cis-Permethrin 100 µg/mL in Acetonitrile, Dr. Ehrenstorfer, LGC Standards
Catalog # 77691-907
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:61949-76-6
- Description:cis-Permethrin 100 µg/mL in Acetonitrile
- Size:1 ml
- Formula:C₂₁H₂₀Cl₂O₃
- Cat. No.:77691-907