(±)-Geosmin 100 µg/mL in Methanol, Dr. Ehrenstorfer, LGC Standards
Catalog # 77691-901
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:16423-19-1
- Description:(±)-Geosmin 100 µg/mL in Methanol
- Size:1 ml
- Formula:C₁₂H₂₂O
- Molecular Weight:182.30 g/mol
- Cat. No.:77691-901