17-beta-Estradiol 100 µg/mL in Acetonitrile, Dr. Ehrenstorfer, LGC Standards
77691-900
New Product
:
- CAS No.:50-28-2
- Description:17-beta-Estradiol 100 µg/mL in Acetonitrile
- Size:1 ml
- Formula:C₁₈H₂₄O₂
- Molecular Weight:272.39 g/mol
- Cat. No.:77691-900