2,4-D 13C6 100 µg/mL in Acetone, Dr. Ehrenstorfer, LGC Standards
Catalog # 77691-879
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:150907-52-1
- Description:2,4-D 13C6 100 µg/mL in Acetone
- Size:1 ml
- Formula:Cl*C₆H₃(Cl)OCH₂COOH
- Molecular Weight:226.99 g/mol
- Cat. No.:77691-879